Learn More
Medchemexpress LLC Eflornithine hydrochloride | 68278-23-9 | 99.9% | 500 MG
SDP
Catalog No. 5000405261
Encompass_Preferred
Eflornithine hydrochloride is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used as a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women.
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Treats African trypanosomiasis
  • Reduces excessive facial hair growth in women
  • Shown to normalize contractile intensity in hypertensive rats
Catalog No. 50-004-05261 Supplier Medchemexpress LLC Supplier No. HYB0744A500MG
Eflornithine hydrochloride is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used as a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women.
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Treats African trypanosomiasis
  • Reduces excessive facial hair growth in women
  • Shown to normalize contractile intensity in hypertensive rats