Learn More
TARGETMOL CHEMICALS INC Eflornithine hydrochloride hyd
SDP
Catalog No. 502876364
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific and irreversible inhibitor of the ornithine decarboxylase (ODC). Eflornithine hydrochloride hydrate has antitumor activity and has also been used in the hirsutism Eflornithine hydrochloride hydrate has antitumor activity and is also used in the treatment of hirsutism. Purity 99%
Catalog No. 50-287-6364 Supplier TARGETMOL CHEMICALS INC Supplier No. T2593500MG
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific and irreversible inhibitor of the ornithine decarboxylase (ODC). Eflornithine hydrochloride hydrate has antitumor activity and has also been used in the hirsutism Eflornithine hydrochloride hydrate has antitumor activity and is also used in the treatment of hirsutism. Purity 99%