Learn More
Medchemexpress LLC Eflornithine (hydrochloride hydrate) | 96020-91-6 | 1 G
SDP
Catalog No. 5000405373
Encompass_Preferred
Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used as a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women. This product also serves as an analytical standard for research and analytical applications.
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Used for treatment of African trypanosomiasis
  • Used for excessive facial hair growth in women
  • Functions as an analytical standard for research applications
Catalog No. 50-004-05373 Supplier Medchemexpress LLC Supplier No. HYB0744B1G
Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used as a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women. This product also serves as an analytical standard for research and analytical applications.
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Used for treatment of African trypanosomiasis
  • Used for excessive facial hair growth in women
  • Functions as an analytical standard for research applications