Learn More
Medchemexpress LLC Eflornithine (hydrochloride hydrate) | 96020-91-6 | 5 G
SDP
Catalog No. 5000408966
Encompass_Preferred
Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used in the treatment of African trypanosomiasis and excessive facial hair growth in women. This product is intended for research and analytical applications.
  • For research and analytical applications
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Recommended storage for powder: 4°C, sealed, away from moisture
  • Recommended storage for in solvent: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture
  • Shipping at room temperature if less than 2 weeks
Catalog No. 50-004-08966 Supplier Medchemexpress LLC Supplier No. HYB0744B5G
Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. It is used in the treatment of African trypanosomiasis and excessive facial hair growth in women. This product is intended for research and analytical applications.
  • For research and analytical applications
  • Specific, irreversible inhibitor of ornithine decarboxylase
  • Recommended storage for powder: 4°C, sealed, away from moisture
  • Recommended storage for in solvent: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture
  • Shipping at room temperature if less than 2 weeks