Learn More
Enzo Life Sciences EFLUXX-ID Green multidrug resistance assay kit (1Kit)
SDP
Catalog No. 502012595
Encompass_Preferred
The EFLUXX-ID Multidrug Resistance Assay Kit allows for functional detection of all three clinically relevant ABC transporter proteins: MDR1 (p-glycoprotein), MRP1/2, and BCRP. The assay uses a hydrophobic, non-fluorescent compound that readily penetrates the cell membrane, where it is hydrolyzed to a hydrophilic fluorescent dye by intracellular esterases. Unless the EFLUXX-ID dye is pumped out of the cell, the esterase cleaved dye is trapped inside the cell. Thus, cells exhibiting drug resistance will have diminished fluorescence. EFLUXX-ID assay is the only available kit for the simultaneous monitoring of all three major ABC reporter proteins with the ability to profile individual pump activity. Applications: Flow Cytometry, Fluorescence microscopy .
Catalog No. 50-201-2595 Supplier Enzo Life Sciences Supplier No. ENZ51029K100
The EFLUXX-ID Multidrug Resistance Assay Kit allows for functional detection of all three clinically relevant ABC transporter proteins: MDR1 (p-glycoprotein), MRP1/2, and BCRP. The assay uses a hydrophobic, non-fluorescent compound that readily penetrates the cell membrane, where it is hydrolyzed to a hydrophilic fluorescent dye by intracellular esterases. Unless the EFLUXX-ID dye is pumped out of the cell, the esterase cleaved dye is trapped inside the cell. Thus, cells exhibiting drug resistance will have diminished fluorescence. EFLUXX-ID assay is the only available kit for the simultaneous monitoring of all three major ABC reporter proteins with the ability to profile individual pump activity. Applications: Flow Cytometry, Fluorescence microscopy .