Learn More
Aviva Systems Biology Corp EFNA4 Antibody - N-terminal region (ARP61596_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61596_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFNA4. It was validated on Western Blot. . Alternate names: EFL4, EPLG4, LERK4, MGC125826ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-416