Learn More
Aviva Systems Biology Corp EFNA5 Antibody - N-terminal region (ARP65430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFNA5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human EFNA5. Alternate names: AF1, EFL5, EPLG7, GLC1M, LERK7, RAGSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-497