Learn More
ABclonal Technology EFNB1 Rabbit pAb
SDP
Catalog No. 5000107562
Encompass_Preferred
The protein encoded by this gene is a type I membrane protein and a ligand of Eph-related receptor tyrosine kinases. It may play a role in cell adhesion and function in the development or maintenance of the nervous system.
Catalog No. 50-001-07562 Supplier ABclonal Technology Supplier No. A145621000UL
The protein encoded by this gene is a type I membrane protein and a ligand of Eph-related receptor tyrosine kinases. It may play a role in cell adhesion and function in the development or maintenance of the nervous system.