Learn More
Cayman Chemical Efonidipine, Purity: ≥98%, CAS: 111011-63-3, 10 mg
SDP
Catalog No. 502946720
Encompass_Preferred
An L- and T-type calcium channel inhibitor; inhibits hamster Cav1.2a and human Cav3.2 (IC50s = 1.8 and 350 nM, respectively); inhibits L- and T-type calcium channels and fetal bovine serum-induced hypertrophy in isolated mouse cardiomyocytes; increases survival in a mouse model of acute myocardial infarction induced by ligation of the left coronary artery at 200 mg/kg; inhibits SARS-CoV-2 Mpro (IC50 = 38.5 µM)
Catalog No. 50-294-6720 Supplier Cayman Chemical Supplier No. 3311410 MG
An L- and T-type calcium channel inhibitor; inhibits hamster Cav1.2a and human Cav3.2 (IC50s = 1.8 and 350 nM, respectively); inhibits L- and T-type calcium channels and fetal bovine serum-induced hypertrophy in isolated mouse cardiomyocytes; increases survival in a mouse model of acute myocardial infarction induced by ligation of the left coronary artery at 200 mg/kg; inhibits SARS-CoV-2 Mpro (IC50 = 38.5 µM)