Learn More
Selleck Chemical LLC Efonidipine HCl ethanolate
SDP
Catalog No. 5000003959
Encompass_Preferred
Efonidipine (NZ-105) hydrochloride monoethanolate is an L- and T-type calcium channel blocker leading to vasodilation and decreased automaticity of the heart Efonidipine hydrochloride monoethanolate also suppresses aldosterone secretion from the adrenal
Catalog No. 50-000-03959 Supplier Selleck Chemical LLC Supplier No. S58985MG
Add to Cart
Add to Cart
Efonidipine (NZ-105) hydrochloride monoethanolate is an L- and T-type calcium channel blocker leading to vasodilation and decreased automaticity of the heart Efonidipine hydrochloride monoethanolate also suppresses aldosterone secretion from the adrenal