Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Efonidipine hydrochloride monoethanolate | 111011-76-8 | 99.8% | 714.18 | C36H45ClN3O8P | 100MG
SDP

Catalog No. 5000237315
Encompass_Preferred

Efonidipine hydrochloride monoethanolate is the monoethanolate hydrochloride salt of efonidipine, a dihydropyridine calcium channel blocker active at both L-type and T-type channels. It is provided as a solid powder or as a DMSO solution for laboratory research and is labeled for research use only.

  • Dual L- and T-type calcium channel blocker.
  • Available as solid powder and 10 mM solution in DMSO.
  • High purity (>99.8%).
  • CAS 111011-76-8 for unambiguous identification.
  • Molecular weight 714.18 g/mol; formula C36H45ClN3O8P.
  • COA and SDS documentation available.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-37315 Supplier Medchemexpress LLC Supplier No. HY12502A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Efonidipine hydrochloride monoethanolate is the monoethanolate hydrochloride salt of efonidipine, a dihydropyridine calcium channel blocker active at both L-type and T-type channels. It is provided as a solid powder or as a DMSO solution for laboratory research and is labeled for research use only.

  • Dual L- and T-type calcium channel blocker.
  • Available as solid powder and 10 mM solution in DMSO.
  • High purity (>99.8%).
  • CAS 111011-76-8 for unambiguous identification.
  • Molecular weight 714.18 g/mol; formula C36H45ClN3O8P.
  • COA and SDS documentation available.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.