Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Efonidipine hydrochloride monoethanolate | 111011-76-8 | MFCD00875717 | >98.0% | 714.18 g/mol | C34H38N3O7P · HCl · C2H5OH | 10MM 1ML
SDP

Catalog No. 5000237345
Encompass_Preferred

Efonidipine hydrochloride monoethanolate is the monoethanolate salt of efonidipine, a dual T-type and L-type calcium channel blocker commonly used in pharmacological research. It is supplied as both a solid and a ready-to-use 10 mM solution in DMSO for in vitro assays and mechanism-of-action studies.

  • Salt form: hydrochloride monoethanolate.
  • CAS registry number: 111011-76-8.
  • Available formats: solid (multiple milligram quantities) and solution (10 mM in DMSO).
  • Purity: >98.0% (HPLC).
  • Molecular weight: 714.18 g/mol (monoethanolate).
  • Chemical formula: C34H38N3O7P · HCl · C2H5OH.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-37345 Supplier Medchemexpress LLC Supplier No. HY12502A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Efonidipine hydrochloride monoethanolate is the monoethanolate salt of efonidipine, a dual T-type and L-type calcium channel blocker commonly used in pharmacological research. It is supplied as both a solid and a ready-to-use 10 mM solution in DMSO for in vitro assays and mechanism-of-action studies.

  • Salt form: hydrochloride monoethanolate.
  • CAS registry number: 111011-76-8.
  • Available formats: solid (multiple milligram quantities) and solution (10 mM in DMSO).
  • Purity: >98.0% (HPLC).
  • Molecular weight: 714.18 g/mol (monoethanolate).
  • Chemical formula: C34H38N3O7P · HCl · C2H5OH.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.