Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Efonidipine hydrochloride monoethanolate | 111011-76-8 | MFCD00875717 | >98.0% | 714.18 g/mol | C36H45ClN3O8P | 25MG
SDP

Catalog No. 5000235293
Encompass_Preferred

Efonidipine hydrochloride monoethanolate is the monoethanolate salt of efonidipine, a dual L-type and T-type calcium channel blocker supplied as a research-grade chemical standard for in vitro pharmacology and biochemical studies. It is intended for research use only.

  • Dual L-type and T-type calcium channel blocker.
  • Research-grade chemical standard for assay and pharmacology studies.
  • High purity (≥98% by HPLC).
  • Available in multiple pack sizes, including 25 mg.
  • Molecular formula C36H45ClN3O8P, molecular weight 714.18 g/mol.
  • Not for human or diagnostic use.

Catalog No. 50-002-35293 Supplier Medchemexpress LLC Supplier No. HY12502A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Efonidipine hydrochloride monoethanolate is the monoethanolate salt of efonidipine, a dual L-type and T-type calcium channel blocker supplied as a research-grade chemical standard for in vitro pharmacology and biochemical studies. It is intended for research use only.

  • Dual L-type and T-type calcium channel blocker.
  • Research-grade chemical standard for assay and pharmacology studies.
  • High purity (≥98% by HPLC).
  • Available in multiple pack sizes, including 25 mg.
  • Molecular formula C36H45ClN3O8P, molecular weight 714.18 g/mol.
  • Not for human or diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.