Learn More
Selleck Chemical LLC Tomivosertib (eFT-508) 2mg 1849590-01-7
SDP
Catalog No. 501365879
Encompass_Preferred
Tomivosertib (eFT-508) is a potent and selective MNK1/2 inhibitor with IC50s of 2.4 nM and 1 nM, respectively. It potentially results in decreased tumor cell proliferation and tumor growth. Tomivosertib (eFT-508) inhibits eIF4E phosphorylation and dramatically downregulates PD-L1 protein abundance. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5879 Supplier Selleck Chemical LLC Supplier No. S82752MG
Tomivosertib (eFT-508) is a potent and selective MNK1/2 inhibitor with IC50s of 2.4 nM and 1 nM, respectively. It potentially results in decreased tumor cell proliferation and tumor growth. Tomivosertib (eFT-508) inhibits eIF4E phosphorylation and dramatically downregulates PD-L1 protein abundance. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.