Learn More
Selleck Chemical LLC EG00229-E1119-5MG
Supplier: Selleck Chemical LLC E11195MG
EG00229 is a neuropilin 1 (NRP1) receptor antagonist with an IC50 of 3 M against VEGF-A binding to NRP1 b1 domain