Learn More
Medchemexpress LLC HY-10799 5mg Medchemexpress, EG00229 CAS:1210945-69-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107995MG
Encompass_Preferred
Medchemexpress, HY-10799 5mg EG00229 CAS:1210945-69-9 EG00229 is a neuropilin 1 (NRP1) receptor antagonist. EG00229 selectively inhibits VEGF-A binding to NRP1 b1 domain with an IC50 of 3 μM, but has no effect on VEGFA binding to VEGFR-1 and VEGFR-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8493