Learn More
Medchemexpress LLC EG00229 trifluoroacetate | 1210945-69-9 | >98.0% | 611.6 g·mol⁻¹ | C17H19N7O5S3 · CF3CO2H | 1 ML
SDP
Catalog No. 5000241833
Encompass_Preferred
EG00229 (trifluoroacetate) is a small-molecule antagonist of neuropilin-1 (NRP1) that blocks VEGF-A interaction with the NRP1 b1/CendR binding pocket. It is used as a research reagent to probe NRP1-dependent signaling, cell migration, and related in vitro biological effects.
Catalog No. 50-002-41833 Supplier Medchemexpress LLC Supplier No. HY1079910MM 1ML
EG00229 (trifluoroacetate) is a small-molecule antagonist of neuropilin-1 (NRP1) that blocks VEGF-A interaction with the NRP1 b1/CendR binding pocket. It is used as a research reagent to probe NRP1-dependent signaling, cell migration, and related in vitro biological effects.