Learn More
Medchemexpress LLC Eg00229 trifluoroacetate | 1210945-69-9 | 98.0% | 611.60 | C19H20F3N7O7S3 | 50 MG
SDP
Catalog No. 5000242954
Encompass_Preferred
EG00229 trifluoroacetate is a small-molecule antagonist of neuropilin-1 (NRP1) that selectively inhibits VEGF-A binding to the NRP1 b1 domain (reported IC50 = 3 μM). Supplied as the trifluoroacetate salt for research use, it is used in studies of VEGF/NRP1 signaling, receptor binding, and cellular assays.
  • Selectively inhibits VEGF-A binding to the NRP1 b1 domain (reported IC50 = 3 μM).
  • Supplied as the trifluoroacetate salt to aid solubility and handling in research workflows.
  • High purity suitable for biochemical and cellular assays.
  • Characterized molecular weight and formula for accurate experimental planning.
  • Available in multiple pack sizes to accommodate varied research needs.
Catalog No. 50-002-42954 Supplier Medchemexpress LLC Supplier No. HY1079950MG
EG00229 trifluoroacetate is a small-molecule antagonist of neuropilin-1 (NRP1) that selectively inhibits VEGF-A binding to the NRP1 b1 domain (reported IC50 = 3 μM). Supplied as the trifluoroacetate salt for research use, it is used in studies of VEGF/NRP1 signaling, receptor binding, and cellular assays.
  • Selectively inhibits VEGF-A binding to the NRP1 b1 domain (reported IC50 = 3 μM).
  • Supplied as the trifluoroacetate salt to aid solubility and handling in research workflows.
  • High purity suitable for biochemical and cellular assays.
  • Characterized molecular weight and formula for accurate experimental planning.
  • Available in multiple pack sizes to accommodate varied research needs.