Learn More
TARGETMOL CHEMICALS INC EG01377 2HCL 5MG
SDP
Catalog No. 502581561
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EG01377 2HCl is a potent bioavailable and selective inhibitor of neuropilin-1 (NRP1) with a Kd value of 1.32 uM and an IC50 value of 609 nM for both EG01377 2HCl against NRP1-a1 and NRP1-b1.EG01377 exhibits anti-angiogenic antimigratory and antitumor activities. purity: 99%
Catalog No. 50-258-1561 Supplier TARGETMOL CHEMICALS INC Supplier No. T11154L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EG01377 2HCl is a potent bioavailable and selective inhibitor of neuropilin-1 (NRP1) with a Kd value of 1.32 uM and an IC50 value of 609 nM for both EG01377 2HCl against NRP1-a1 and NRP1-b1.EG01377 exhibits anti-angiogenic antimigratory and antitumor activities. purity: 99%