Learn More
Medchemexpress LLC EG01377 dihydrochloride | 2749438-61-5 | 99.0% | 659.60 | 1 ML
SDP
Catalog No. 5000435628
Encompass_Preferred
EG01377 dihydrochloride is a potent, bioavailable, and selective inhibitor of neuropilin-1 (NRP1), with a Kd of 1.32 μM, and IC50s of 609 nM for both NRP1-a1 and NRP1-b1. It exhibits antiangiogenic, antimigratory, and antitumor effects.
  • Inhibits vascular endothelial growth factor A (VEGF-A) stimulated tyrosine phosphorylation of VEGF-R2/KDR.
  • Reduces HUVEC cell migration in response to VEGFA.
  • Can delay VEGF-induced wound closure.
  • Reduces network area, length, and branching points.
  • Reduces VEGF-induced angiogenesis.
  • Reduces A375P (malignant melanoma) spheroid outgrowth in combination with VEGFA.
  • Blocks the production of transforming growth factor beta (TGFβ) by Nrp1+ regulatory T-cell SMAD3/AKT (Tregs) in the presence of tumor cell-derived factors.
  • Exhibits an encouraging half-life sufficient to sustain once per day dosing in mice.
Catalog No. 50-004-35628 Supplier Medchemexpress LLC Supplier No. HY112151A10MM/1ML
EG01377 dihydrochloride is a potent, bioavailable, and selective inhibitor of neuropilin-1 (NRP1), with a Kd of 1.32 μM, and IC50s of 609 nM for both NRP1-a1 and NRP1-b1. It exhibits antiangiogenic, antimigratory, and antitumor effects.
  • Inhibits vascular endothelial growth factor A (VEGF-A) stimulated tyrosine phosphorylation of VEGF-R2/KDR.
  • Reduces HUVEC cell migration in response to VEGFA.
  • Can delay VEGF-induced wound closure.
  • Reduces network area, length, and branching points.
  • Reduces VEGF-induced angiogenesis.
  • Reduces A375P (malignant melanoma) spheroid outgrowth in combination with VEGFA.
  • Blocks the production of transforming growth factor beta (TGFβ) by Nrp1+ regulatory T-cell SMAD3/AKT (Tregs) in the presence of tumor cell-derived factors.
  • Exhibits an encouraging half-life sufficient to sustain once per day dosing in mice.