Learn More
Labtech Inc EG35APlus hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 5.0 C, Power supply: 230V, 50/60Hz
Catalog No. 5000496187
Encompass_Preferred
EG35APlus hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 5.0 C, Power supply: 230V, 50/60Hz
Catalog No. 50-004-96187 Supplier Labtech Inc Supplier No. LH2081
EG35APlus hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 5.0 C, Power supply: 230V, 50/60Hz