Learn More
Labtech Inc EG35B hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 2.0 C, Power supply: 230V, 50/60Hz
Catalog No. 5000496188
Encompass_Preferred
EG35B hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 2.0 C, Power supply: 230V, 50/60Hz
Catalog No. 50-004-96188 Supplier Labtech Inc Supplier No. LH2090
EG35B hotplate, Heating surface: Stainless steel, Temperature range: Ambient - 350℃, Heating power: 3000W, Temperature Accuracy: +/- 2.0 C, Power supply: 230V, 50/60Hz