Learn More
Labtech Inc EG37APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496180
Encompass_Preferred
EG37APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96180 Supplier Labtech Inc Supplier No. LH2201
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
EG37APlus hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 5.0 C, Power Supply: 230V, 50/60Hz