Learn More
Labtech Inc EG37B hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496196
Encompass_Preferred
EG37B hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96196 Supplier Labtech Inc Supplier No. LH2210
EG37B hotplate, Heating surface: Graphite, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz