Learn More
Labtech Inc EG37C hotplate, Heating surface: Graphite with PTFE coating, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 5000496176
Encompass_Preferred
EG37C hotplate, Heating surface: Graphite with PTFE coating, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz
Catalog No. 50-004-96176 Supplier Labtech Inc Supplier No. LH2211
EG37C hotplate, Heating surface: Graphite with PTFE coating, Temperature range: Ambient - 370℃, Heating power: 3200W, Temperature Accuracy: +/- 2.0 C, Power Supply: 230V, 50/60Hz