Learn More
Aviva Systems Biology Corp EG546729 Antibody - C-terminal region (ARP44397_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44397_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EG546729. It was validated on Western Blot. . Alternate names: EG546729ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-143