Learn More
ATCC E.G7-OVA; Lymphoma; Mouse (Mus musculus)
Catalog No. 502383242
Encompass
E.G7-OVA [derivative of EL4] T lymphoblasts are T lymphoblasts derived in 1988 from the C57BL/6 (H-2 b) mouse lymphoma cell line EL4 (ATCC TIB-39). E.G7-OVA [derivative of EL4] is a model system for studying major histocompatibility complex (MHC) class I restricted responses of cytotoxic T lymphocytes in mice.
Catalog No. 50-238-3242 Supplier ATCC Supplier No. CRL2113
E.G7-OVA [derivative of EL4] T lymphoblasts are T lymphoblasts derived in 1988 from the C57BL/6 (H-2 b) mouse lymphoma cell line EL4 (ATCC TIB-39). E.G7-OVA [derivative of EL4] is a model system for studying major histocompatibility complex (MHC) class I restricted responses of cytotoxic T lymphocytes in mice.