Learn More
TA Instruments EGA Furnace TGA 55, TGA 550, Low-Volume Vacuum-Tight Furnace, Evolved Gas Testing, FT-IR, MS, GC, Quartz Tube, 20 ML, Swagelok Fitting, EGA Furnace TGA 55
Catalog No. 5000580737 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments EGA Furnace for the TGA 55 and TGA 550 is a low-volume and vacuum-tight furnace designed for evolved gas testing with FT-IR, MS, and GC. Furnace windings are outside the quartz tube, and the reduced 20 mL volume prevents dilution of off-gas concentration. The furnace connects via a standard 1/8 inch Swagelok fitting.
Catalog No. 50-005-80737 Supplier TA Instruments Supplier No. 956075.901
Waters TA Instruments EGA Furnace for the TGA 55 and TGA 550 is a low-volume and vacuum-tight furnace designed for evolved gas testing with FT-IR, MS, and GC. Furnace windings are outside the quartz tube, and the reduced 20 mL volume prevents dilution of off-gas concentration. The furnace connects via a standard 1/8 inch Swagelok fitting.