Learn More
TA Instruments EGA Gas Sampling Device FTIR 230V 50Hz, Inert Silcosteel Transfer Tube, Gas Flow-Controlled Pump System, Trigger Cable, Touch Screen Controller, Requires Separately Sold Adapter and FTIR Gas Cell
Catalog No. 5000580815 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments EGA Gas Sampling Device for FTIR provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to an FTIR for gas analysis. It features an inert, corrosion-resistant, non-reactive Silcosteel transfer tube that can be heated to 350 °C, a gas flow-controlled pump system for aspiration, filtration, and thermal stabilization of evolved gases, a trigger cable for FTIR, and a touch screen controller. It requires a separately sold EGA Interface gas sampling adapter and FTIR gas cell for operation.
Catalog No. 50-005-80815 Supplier TA Instruments Supplier No. 206405.902
Waters TA Instruments EGA Gas Sampling Device for FTIR provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to an FTIR for gas analysis. It features an inert, corrosion-resistant, non-reactive Silcosteel transfer tube that can be heated to 350 °C, a gas flow-controlled pump system for aspiration, filtration, and thermal stabilization of evolved gases, a trigger cable for FTIR, and a touch screen controller. It requires a separately sold EGA Interface gas sampling adapter and FTIR gas cell for operation.