Learn More
TA Instruments EGA Gas Sampling Device FTIR GCMS 110V 60Hz, Inert Silcosteel Transfer Tube, Gas Flow-Controlled Pump System, Trigger Cables, Touch Screen Controller, Requires Separately Sold Adapter, FTIR Gas Cell, GCMS Gas Valve Injection Block
Catalog No. 5000580771 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments EGA Gas Sampling Device for FTIR and GCMS provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to both FTIR and GCMS for gas analysis. It includes an inert, corrosion-resistant, non-reactive Silcosteel transfer tube heated to 350 °C, a gas flow-controlled pump system for aspiration, filtration, and thermal stabilization of evolved gases, trigger cables for FTIR and GCMS, and a touch screen controller. It requires separately sold EGA Interface gas sampling adapter, FTIR gas cell, and GCMS gas valve injection block.
Catalog No. 50-005-80771 Supplier TA Instruments Supplier No. 206407.901
Waters TA Instruments EGA Gas Sampling Device for FTIR and GCMS provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to both FTIR and GCMS for gas analysis. It includes an inert, corrosion-resistant, non-reactive Silcosteel transfer tube heated to 350 °C, a gas flow-controlled pump system for aspiration, filtration, and thermal stabilization of evolved gases, trigger cables for FTIR and GCMS, and a touch screen controller. It requires separately sold EGA Interface gas sampling adapter, FTIR gas cell, and GCMS gas valve injection block.