Learn More
TA Instruments EGA Gas Sampling Device GCMS 110V 60Hz, Inert Silcosteel Transfer Tube, Trigger Cable, Touch Screen Controller, Requires Separately Sold Adapter and GCMS Gas Valve Injection Block
Catalog No. 5000580770 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments EGA Gas Sampling Device for GCMS provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to a GCMS for gas analysis. It includes an inert, corrosion-resistant, non-reactive Silcosteel transfer tube heated to 350 °C, a trigger cable for GCMS, and a touch screen controller. It requires a separately sold EGA Interface gas sampling adapter and GCMS gas valve injection block for operation.
Catalog No. 50-005-80770 Supplier TA Instruments Supplier No. 206406.901
Waters TA Instruments EGA Gas Sampling Device for GCMS provides contamination-free transfer of evolved gas from a Discovery TGA/Discovery SDT to a GCMS for gas analysis. It includes an inert, corrosion-resistant, non-reactive Silcosteel transfer tube heated to 350 °C, a trigger cable for GCMS, and a touch screen controller. It requires a separately sold EGA Interface gas sampling adapter and GCMS gas valve injection block for operation.