Learn More
TA Instruments EGA Interface Gas Sampling Adapter, Heated Adapter for Connection to Discovery TGA 55/550 with EGA Furnace, for Transfer of Evolved Gas to FTIR or GCMS, Sellable Only with Separately Sold EGA Gas Sampling Device
Catalog No. 5000580794 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments EGA Interface Gas Sampling Adapter is a heated adapter for connecting a Discovery TGA 55 or 550 equipped with an EGA furnace to a separately sold EGA Gas Sampling Device. It allows transfer of evolved gas to external analysis instruments such as FTIR or GCMS. It connects only to TGA 55 or 550 with EGA furnace and is functional only with a separately sold EGA Gas Sampling Device.
Catalog No. 50-005-80794 Supplier TA Instruments Supplier No. 206411.001
Waters TA Instruments EGA Interface Gas Sampling Adapter is a heated adapter for connecting a Discovery TGA 55 or 550 equipped with an EGA furnace to a separately sold EGA Gas Sampling Device. It allows transfer of evolved gas to external analysis instruments such as FTIR or GCMS. It connects only to TGA 55 or 550 with EGA furnace and is functional only with a separately sold EGA Gas Sampling Device.