Learn More
Selleck Chemical LLC Eganelisib
SDP
Catalog No. 5000001556
Encompass_Preferred
Eganelisib (IPI-549) is a potent inhibitor of PI3K- with 100-fold selectivity over other lipid and protein kinases The biochemical IC50 for PI3K- is 16 nM
Catalog No. 50-000-01556 Supplier Selleck Chemical LLC Supplier No. S8330100MG
Eganelisib (IPI-549) is a potent inhibitor of PI3K- with 100-fold selectivity over other lipid and protein kinases The biochemical IC50 for PI3K- is 16 nM