Learn More
Medchemexpress LLC Eganelisib (IPI-549) | 1693758-51-8 | MFCD30533720 | 99.7% | 528.56 g·mol⁻¹ | C30H24N8O2 | 10 MG
SDP
Catalog No. 5000215890
Encompass_Preferred
Eganelisib (IPI-549) is an orally bioavailable, selective small-molecule inhibitor of the gamma isoform of phosphoinositide 3-kinase (PI3Kγ) used in research and preclinical immuno-oncology studies. It demonstrates potent PI3Kγ inhibition (IC50 ≈ 16 nM) and >100-fold selectivity over other lipid and protein kinases.
  • Potent PI3Kγ inhibition with IC50 ≈ 16 nM.
  • Greater than 100-fold selectivity over other lipid and protein kinases.
  • High reported purity suitable for research applications.
  • Known molecular weight and CAS for unambiguous identification.
  • Supplied in small quantities appropriate for preclinical studies.
Catalog No. 50-002-15890 Supplier Medchemexpress LLC Supplier No. HY10071610MG
Eganelisib (IPI-549) is an orally bioavailable, selective small-molecule inhibitor of the gamma isoform of phosphoinositide 3-kinase (PI3Kγ) used in research and preclinical immuno-oncology studies. It demonstrates potent PI3Kγ inhibition (IC50 ≈ 16 nM) and >100-fold selectivity over other lipid and protein kinases.
  • Potent PI3Kγ inhibition with IC50 ≈ 16 nM.
  • Greater than 100-fold selectivity over other lipid and protein kinases.
  • High reported purity suitable for research applications.
  • Known molecular weight and CAS for unambiguous identification.
  • Supplied in small quantities appropriate for preclinical studies.