Learn More
New England Biolabs, Inc. Endoglycoceramidase I (EGCase I) – 150 milliunits
SDP
Supplier:  New England Biolabs, Inc. P0773S
Encompass
Endoglycoceramidase I (EGCase I) catalyzes the hydrolysis of the b-glycosidic linkage between oligosaccharides and ceramides in various glycosphingolipids. The enzyme leaves the oligosaccharide and ceramide intact, keeping them suitable for further analysis.
Catalog No. 50-202-4050