Learn More
Thermo Scientific EGC500 K2CO3 CAR
Catalog No. NC0789926 Shop All Thermo Scientific Products
Encompass
Prevent baseline shift, increase sensitivity, improve resolution, and ensure consistent peak integration by using eluent generation. A range of Thermo Scientific™ Dionex™ EGC Cartridges are available for the production of hydroxide, carbonate, bicarbonate and methanesulfonic acid eluents. Eluent generation eliminates the need to handle acids and bases, which are traditionally required for the preparation of IC eluents, and allows chromatographers to run a full range of gradient and isocratic separations more effectively than hand-made eluents.
This item is not returnable. View return policy
Catalog No. NC0789926 Supplier Thermo Scientific Supplier No. 088453
Prevent baseline shift, increase sensitivity, improve resolution, and ensure consistent peak integration by using eluent generation. A range of Thermo Scientific™ Dionex™ EGC Cartridges are available for the production of hydroxide, carbonate, bicarbonate and methanesulfonic acid eluents. Eluent generation eliminates the need to handle acids and bases, which are traditionally required for the preparation of IC eluents, and allows chromatographers to run a full range of gradient and isocratic separations more effectively than hand-made eluents.