Learn More
Medchemexpress LLC Egcg octaacetate | 148707-39-5 | 1 MG
SDP
Catalog No. 5000420134
Encompass_Preferred
EGCG Octaacetate (AcEGCG) is a proagent of Green tea epigallocatechin-3-gallate (EGCG). It works by down-regulating PI3K/Akt/NFκB phosphorylation and p65 acetylation, offering a range of biological activities including antibacterial effects.
  • Decreases proinflammatory mediator levels.
  • Functions as an antibacterial compound for gram-positive bacteria (GPB) and gram-negative bacteria (GNB).
  • Exhibits antioxidant, anti-angiogenesis, anti-inflammatory, and antitumor properties.
  • Reduces colitis-driven colon cancer in mouse models.
Catalog No. 50-004-20134 Supplier Medchemexpress LLC Supplier No. HYN62631MG
EGCG Octaacetate (AcEGCG) is a proagent of Green tea epigallocatechin-3-gallate (EGCG). It works by down-regulating PI3K/Akt/NFκB phosphorylation and p65 acetylation, offering a range of biological activities including antibacterial effects.
  • Decreases proinflammatory mediator levels.
  • Functions as an antibacterial compound for gram-positive bacteria (GPB) and gram-negative bacteria (GNB).
  • Exhibits antioxidant, anti-angiogenesis, anti-inflammatory, and antitumor properties.
  • Reduces colitis-driven colon cancer in mouse models.