Learn More
Medchemexpress LLC HY-P7092A 50ug Medchemexpress, EGF (CHO-expressed), Rat CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP7092A50UG
Encompass_Preferred
Medchemexpress, HY-P7092A 50ug EGF (CHO-expressed), Rat CAS: Recombinant Rat Epidermal Growth Factor (CHO-expressed) is a potent epidermal growth factor, promotes epithelial proliferation and migration, and increases epithelial wound closure and shortens healing time. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3107