Learn More
Sino Biological EGF / Epidermal Growth Factor Antibody, Rabbit PAb 100uL
Catalog No. 502248216
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Canine EGF / Epidermal Growth Factor (Catalog#70013-D07E; Q9BEA0; Asn973-Arg1024). Total IgG was purified by Protein A affinity chromatography.
This item is not returnable. View return policy
Catalog No. 50-224-8216 Supplier Sino Biological Supplier No. 70013RP01100
Produced in rabbits immunized with purified, recombinant Canine EGF / Epidermal Growth Factor (Catalog#70013-D07E; Q9BEA0; Asn973-Arg1024). Total IgG was purified by Protein A affinity chromatography.