Learn More
Sino Biological Anti-EGF Antibody, Mouse Monoclonal
Supplier:  Sino Biological 70013MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Canine EGF / Epidermal Growth Factor ( Catalog#70013-D07E; Q9BEA0; Asn973-Arg1024). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8212