Learn More
ACROBiosystems ACROBiosystems Mouse EGF Protein, Fc Tag
Supplier:  ACROBiosystems EGFM5265100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse EGF, Fc Tag (EGF-M5265) is expressed from human 293 cells (HEK293). It contains AA Asn 977 - Arg 1029 (Accession # P01132-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 32.5 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse EGF R, His Tag (Cat. No. EGR-M5224) at 5 μg/mL (100 μL/well) can bind Mouse EGF, Fc Tag (Cat. No. EGF-M5265) with a linear range of 0.2-5 ng/mL (QC tested).;Captured Mouse EGF R, His Tag (Cat. No. EGR-M5224) on CM5 Chip via anti-His antibody can bind Mouse EGF, Fc Tag (Cat. No. EGF-M5265) with an affinity constant of 5.44 nM as determined in SPR assay (Biacore T200 ) (Routinely tested).
Catalog No. 50-195-7005