Learn More
Enzo Life Sciences Epidermal growth factor (human) (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI908060100
Encompass_Preferred
Alternative name: EGF. Purity: ≥98% (SDS-PAGE; RP-HPLC). Formulation: Lyophilized. Sterile filtered liquid from a concentrated (1mg/ml) solution containing PBS, pH 7.4 It is recommend to reconstitute this protein in sterile 18MΩ-cm H2O to not less than 100µg/ml. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: ELISA, WB in vitro Assay.
Catalog No. 50-200-7090