Learn More
Medchemexpress LLC EGF Human HEK293 50ug
SDP
Catalog No. 5000182785
Encompass_Preferred
The Progranulin/PGRN protein is involved in a variety of cellular processes including cell growth inflammation and neurodegeneration It acts as a regulator of tissue repair and has been implicated in the pathogenesis of several diseases including Alzheimer s disease and frontotemporal dementia Progranulin/PGRN interacts with various receptors and signaling pathways to exert its effects on cellular function and disease progression EGF Protein Human (HEK293 N-hFc) is the recombinant human-derived EGF protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-82785 Supplier Medchemexpress LLC Supplier No. HYP72982A50UG
The Progranulin/PGRN protein is involved in a variety of cellular processes including cell growth inflammation and neurodegeneration It acts as a regulator of tissue repair and has been implicated in the pathogenesis of several diseases including Alzheimer s disease and frontotemporal dementia Progranulin/PGRN interacts with various receptors and signaling pathways to exert its effects on cellular function and disease progression EGF Protein Human (HEK293 N-hFc) is the recombinant human-derived EGF protein expressed by HEK293 with N-hFc labeled tag