Learn More
Medchemexpress LLC HY-P7067 100ug Medchemexpress, EGF, Mouse CAS: Purity:>98%
SDP
Catalog No. 501934310
Encompass_Preferred
Medchemexpress, HY-P7067 100ug EGF, Mouse CAS: Recombinant Mouse Epidermal Growth Factor is a small polypeptide that stimulates the proliferation of the epidermis and several epithelial tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4310 Supplier Medchemexpress LLC Supplier No. HYP7067100UG
Medchemexpress, HY-P7067 100ug EGF, Mouse CAS: Recombinant Mouse Epidermal Growth Factor is a small polypeptide that stimulates the proliferation of the epidermis and several epithelial tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.