Learn More
Medchemexpress LLC EGF Mouse 10ug
SDP
Catalog No. 5000184907
Encompass_Preferred
EGF Protein, Mouse is a recombinant epidermal growth factor supplied as a lyophilized, tag-free powder for research applications. It promotes proliferation of epidermal and other epithelial tissues, is produced in E. coli, and has an approximate molecular weight of 6-7 kDa with >95% purity. The product is provided as a 10 μg vial, should be stored frozen at -20°C, and can be aliquoted and frozen at -20°C or -80°C after reconstitution for extended storage.
  • Stimulates epidermal and epithelial cell proliferation.
  • Recombinant, tag free protein produced in E. coli.
  • High purity (>95%) suitable for biochemical assays.
  • Supplied as lyophilized powder for extended shelf life.
  • Provides 10 μg per vial for small-scale experiments.
  • Stable when stored frozen and after reconstitution if aliquoted.
Catalog No. 50-001-84907 Supplier Medchemexpress LLC Supplier No. HYP706710UG
EGF Protein, Mouse is a recombinant epidermal growth factor supplied as a lyophilized, tag-free powder for research applications. It promotes proliferation of epidermal and other epithelial tissues, is produced in E. coli, and has an approximate molecular weight of 6-7 kDa with >95% purity. The product is provided as a 10 μg vial, should be stored frozen at -20°C, and can be aliquoted and frozen at -20°C or -80°C after reconstitution for extended storage.
  • Stimulates epidermal and epithelial cell proliferation.
  • Recombinant, tag free protein produced in E. coli.
  • High purity (>95%) suitable for biochemical assays.
  • Supplied as lyophilized powder for extended shelf life.
  • Provides 10 μg per vial for small-scale experiments.
  • Stable when stored frozen and after reconstitution if aliquoted.