Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0297210
Encompass
Epidermal Growth Factor (EGF) is a potent growth factor that stimulates the proliferation of various epidermal and epithelial cells. Additionally EGF has been shown to inhibit gastric secretion and to be involved in wound healing.
Catalog No. 50-252-9932