Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse EGF ELISA Kit

Catalog No. EMEGFX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMEGFX5 5 x 96 Tests
EMEGF 96 Tests
EMEGFX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EMEGFX5 Supplier Invitrogen™ Supplier No. EMEGFX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Mouse Epidermal Growth Factor (Ms EGF) ELISA quantitates Ms EGF in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms EGF. Principle of the method The Mouse EGF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01132
Assay Range 3.28-800 pg/mL
Assay Sensitivity 3 pg/mL
Calibration Range 3 pg/mL to 800 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Sufficient For 5 x 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; UrogastroneAI790464; beta-urogastrone; EGF; Epidermal growth factor; epidermal growth factor (beta-urogastrone); epidermal growth factor precursor; H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); sb:eu639; URG; Urogastrone; AI790464
Gene ID (Entrez) 13645
Gene Symbol EGF
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases EGF
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Development
Sample Volume Plasma 5 μL, Serum 5 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym AI790464
Target EGF
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.