Learn More
Medchemexpress LLC EGF Mouse N-His 100ug
SDP
Catalog No. 5000184549
Encompass_Preferred
Recombinant mouse epidermal growth factor (EGF) with an N-terminal His tag, provided as 100 μg for in vitro research. EGF binds the epidermal growth factor receptor (EGFR) to stimulate cellular proliferation and support wound-healing responses. Common applications include cell signaling studies, proliferation assays, and tissue-regeneration or organoid models.
  • Activates EGFR-mediated signaling in vitro
  • Promotes epithelial and epidermal cell proliferation
  • Useful for cell proliferation, wound-healing, and organoid assays
  • Expressed in E. coli and carries an N-terminal His tag
  • Supplied as 100 μg suitable for small-scale experiments
Catalog No. 50-001-84549 Supplier Medchemexpress LLC Supplier No. HYP70590A100UG
Recombinant mouse epidermal growth factor (EGF) with an N-terminal His tag, provided as 100 μg for in vitro research. EGF binds the epidermal growth factor receptor (EGFR) to stimulate cellular proliferation and support wound-healing responses. Common applications include cell signaling studies, proliferation assays, and tissue-regeneration or organoid models.
  • Activates EGFR-mediated signaling in vitro
  • Promotes epithelial and epidermal cell proliferation
  • Useful for cell proliferation, wound-healing, and organoid assays
  • Expressed in E. coli and carries an N-terminal His tag
  • Supplied as 100 μg suitable for small-scale experiments