Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EGF Mouse N-His 50ug
SDP

Catalog No. 5000184951
Encompass_Preferred

Recombinant full-length mouse epidermal growth factor (EGF) with an N-terminal hexahistidine tag, expressed in Escherichia coli and supplied lyophilized. The protein activates EGFR-mediated signaling and promotes cell proliferation; it is provided at high purity and intended for cell signaling, proliferation assays, and tissue-repair studies. Supplied as 50 μg vials lyophilized from 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.

  • Full-length mouse EGF (53 amino acids).
  • N-terminal hexahistidine tag for affinity purification and detection.
  • Expressed in Escherichia coli, non-glycosylated.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 9-14 kDa.
  • Lyophilized formulation from 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.
  • Stable when frozen; aliquot and store at -20°C or -80°C for long-term storage.
  • Suitable for cell signaling and proliferation assays.

Catalog No. 50-001-84951 Supplier Medchemexpress LLC Supplier No. HYP70590A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length mouse epidermal growth factor (EGF) with an N-terminal hexahistidine tag, expressed in Escherichia coli and supplied lyophilized. The protein activates EGFR-mediated signaling and promotes cell proliferation; it is provided at high purity and intended for cell signaling, proliferation assays, and tissue-repair studies. Supplied as 50 μg vials lyophilized from 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.

  • Full-length mouse EGF (53 amino acids).
  • N-terminal hexahistidine tag for affinity purification and detection.
  • Expressed in Escherichia coli, non-glycosylated.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Molecular weight approximately 9-14 kDa.
  • Lyophilized formulation from 20 mM Tris-HCl, 150 mM NaCl, pH 8.0.
  • Stable when frozen; aliquot and store at -20°C or -80°C for long-term storage.
  • Suitable for cell signaling and proliferation assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.