Learn More
ACROBiosystems ACROBiosystems Human EGF Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems EGFH525B100UG
Encompass_Preferred
Recombinant Protein;100UG;Human EGF, Mouse IgG2a Fc Tag (EGF-H525b) is expressed from human 293 cells (HEK293). It contains AA Asn 971 - Arg 1023 (Accession # P01133-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 33.1 kDa. The protein migrates as 35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human EGF R, Fc Tag (Cat. No. EGR-H5252) at 5 μg/mL (100 μL/well)can bind Human EGF, Mouse IgG2a Fc Tag (Cat. No. EGF-H525b) with a linear range of 0.3-10 ng/mL (QC tested).
Catalog No. 50-195-7220